accessibility menu, dialog, popup

UserName

Viewing profile as user:
HON

CAS RN 2157-52-0

IUPAC Name: N-(9H-fluoren-9-ylidene)hydroxylamine
Synonyms: fluorenone oxime
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: CRNNFEKVPRFZKJ-UHFFFAOYSA-N
SMILES: ON=C1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)