accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21581-45-3

IUPAC Name: 2-(3,4-dichlorophenyl)ethan-1-aminium
Synonyms: 2-(3,4-dichlorophenyl)ethanaminium
Molecular Weight (g/mol): 191.07
Molecular Formula: C8H10Cl2N
InChi Key: MQPUAVYKVIHUJP-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)