CAS RN 2159-42-4
IUPAC Name:
2-(3,4-dimethylbenzoyl)benzoate
Synonyms:
2-(3,4-dimethylbenzoyl)benzoate
Molecular Weight (g/mol):
253.28
Molecular Formula:
C16H13O3
InChi Key:
AYVFSZDAFPVJOA-UHFFFAOYSA-M
SMILES:
CC1=CC=C(C=C1C)C(=O)C1=CC=CC=C1C([O-])=O
1
–
1
of
1
results
1
3',4'-Dimethylbenzophenone-2-carboxylic Acid 98.0+%, TCI America™
CAS: 2159-42-4 Molecular Formula: C16H14O3 Molecular Weight (g/mol): 254.285 InChI Key: AYVFSZDAFPVJOA-UHFFFAOYSA-N Synonym: 2-(3,4-Dimethylbenzoyl)benzoic Acid PubChem CID: 304868 IUPAC Name: 2-(3,4-dimethylbenzoyl)benzoic acid SMILES: CC1=C(C=C(C=C1)C(=O)C2=CC=CC=C2C(=O)O)C