accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 215928-54-4

IUPAC Name: 5-(thiophen-2-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one
Synonyms: 5-(thiophen-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 234.29
Molecular Formula: C10H6N2OS2
InChi Key: LFUBTAOTPHKKOS-UHFFFAOYSA-N
SMILES: O=C1NC=NC2=C1C(=CS2)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results