accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 215943-79-6

IUPAC Name: (2R,3S,4R,5R)-4-amino-5-(2,6-diamino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Synonyms: (2R,3S,4R,5R)-4-amino-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol): 281.28
Molecular Formula: C10H15N7O3
InChi Key: DIFJLWYUVQLAHS-SJLGBTOWNA-N
SMILES: N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=C(N)N=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results