accessibility menu, dialog, popup

UserName

Viewing profile as user:
SBrN

CAS RN 2160-62-5

IUPAC Name: 5-bromothiophene-2-carbonitrile
Synonyms: 5-bromothiophene-2-carbonitrile
Molecular Weight (g/mol): 188.04
Molecular Formula: C5H2BrNS
InChi Key: YVVHCBNJWHPMCQ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results