accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NOHHO

CAS RN 2160-93-2

IUPAC Name: 2-[tert-butyl(2-hydroxyethyl)amino]ethan-1-ol
Synonyms: tert-butyldiethanolamine
Molecular Weight (g/mol): 161.25
Molecular Formula: C8H19NO2
InChi Key: XHJGXOOOMKCJPP-UHFFFAOYSA-N
SMILES: CC(C)(C)N(CCO)CCO
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)