accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNCO

CAS RN 216059-84-6

IUPAC Name: 2-(4-isocyanatophenyl)pyrimidine
Synonyms: 2-(4-isocyanatophenyl)pyrimidine
Molecular Weight (g/mol): 197.2
Molecular Formula: C11H7N3O
InChi Key: VGGNOZLRRMBBQL-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=C(C=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results