accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOClBrCl

CAS RN 21609-90-5

IUPAC Name: O-4-bromo-2,5-dichlorophenyl O-methyl phenylphosphonothioate
Synonyms: leptophos
Molecular Weight (g/mol): 412.06
Molecular Formula: C13H10BrCl2O2PS
InChi Key: CVRALZAYCYJELZ-UHFFFAOYNA-N
SMILES: COP(=S)(OC1=CC(Cl)=C(Br)C=C1Cl)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results