CAS RN 2161-86-6
IUPAC Name:
1-(3-acetyl-2,4,6-trihydroxyphenyl)ethan-1-one
Synonyms:
2,4-diacetylphloroglucinol
Molecular Weight (g/mol):
210.19
Molecular Formula:
C10H10O5
InChi Key:
PIFFQYJYNWXNGE-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(O)C(C(C)=O)=C(O)C=C1O
1
–
1
of
1
results
1
Promotions Available
2,4-Diacetylphloroglucinol, 97%
2,4-Diacetylphloroglucinol, 97%
CAS: 2161-86-6 Molecular Formula: C10H10O5 Molecular Weight (g/mol): 210.19 MDL Number: MFCD00100483 InChI Key: PIFFQYJYNWXNGE-UHFFFAOYSA-N Synonym: 2,4-diacetylphloroglucinol,1-3-acetyl-2,4,6-trihydroxyphenyl ethan-1-one,diacetylphloroglucinol,phloroglucinol, diacetyl,unii-8xv4yyo3wn,8xv4yyo3wn,1-3-acetyl-2,4,6-trihydroxyphenyl ethanone,1,1'-2,4,6-trihydroxy-1,3-phenylene bisethanone,ethanone,1,1'-2,4,6-trihydroxy-1,3-phenylene bis,1-3-acetyl-2,4,6-trihydroxy-phenyl ethanone PubChem CID: 16547 ChEBI: CHEBI:78688 IUPAC Name: 1-(3-acetyl-2,4,6-trihydroxyphenyl)ethanone SMILES: CC(=O)C1=C(O)C(C(C)=O)=C(O)C=C1O