accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3O

CAS RN 216144-29-5

IUPAC Name: 3-(diethoxymethyl)furan
Synonyms: 3-(diethoxymethyl)furan
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: MTZMXGCDOYBCPN-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=COC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results