accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 216159-03-4

IUPAC Name: 4-bromo-2-fluorobenzene-1-sulfonyl chloride
Synonyms: 4-bromo-2-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 273.5
Molecular Formula: C6H3BrClFO2S
InChi Key: XNYBZLRIUHNRQY-UHFFFAOYSA-N
SMILES: FC1=C(C=CC(Br)=C1)S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)