accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3CCH3NC(?)NCH3CH3H3CCH3

CAS RN 2162-74-5

IUPAC Name: N,N'-bis[2,6-bis(propan-2-yl)phenyl]methanediimine
Synonyms: N,N'-bis(2,6-diisopropylphenyl)methanediimine
Molecular Weight (g/mol): 362.56
Molecular Formula: C25H34N2
InChi Key: XLDBGFGREOMWSL-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N=C=NC1=C(C=CC=C1C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results