accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHHO

CAS RN 2163-42-0

IUPAC Name: 2-methylpropane-1,3-diol
Synonyms: methylpropanediol
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: QWGRWMMWNDWRQN-UHFFFAOYSA-N
SMILES: CC(CO)CO
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)