accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2163-48-6

IUPAC Name: 1,3-diethyl 2-propylpropanedioate
Synonyms: 1,3-diethyl 2-propylpropanedioate
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: GRRSDGHTSMJICM-UHFFFAOYSA-N
SMILES: CCCC(C(=O)OCC)C(=O)OCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)