accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21635-90-5

IUPAC Name: tetracyclo[6.2.1.1³,⁶.0²,⁷]dodec-4-ene
Synonyms: tetracyclo[6.2.1.1³,⁶.0²,⁷]dodec-4-ene
Molecular Weight (g/mol): 160.26
Molecular Formula: C12H16
InChi Key: XBFJAVXCNXDMBH-UHFFFAOYNA-N
SMILES: C1CC2CC1C1C3CC(C=C3)C21
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results