accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFF

CAS RN 216393-55-4

IUPAC Name: methyl 3,5-difluorobenzoate
Synonyms: methyl 3,5-difluorobenzoate
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: JBEJPGWPXIIQBB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results