accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrClSOOCl

CAS RN 216394-05-7

IUPAC Name: 5-bromo-6-chloropyridine-3-sulfonyl chloride
Synonyms: 5-bromo-6-chloropyridine-3-sulfonyl chloride
Molecular Weight (g/mol): 290.94
Molecular Formula: C5H2BrCl2NO2S
InChi Key: TURGMVYIESHZBE-UHFFFAOYSA-N
SMILES: ClC1=C(Br)C=C(C=N1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results