accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21643-42-5

IUPAC Name: tetradecyl prop-2-enoate
Synonyms: tetradecyl prop-2-enoate
Molecular Weight (g/mol): 268.44
Molecular Formula: C17H32O2
InChi Key: XZHNPVKXBNDGJD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCOC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results