accessibility menu, dialog, popup

UserName

Viewing profile as user:
BCH3FFFK

CAS RN 216434-82-1

IUPAC Name: potassium trifluoro(4-methylphenyl)boranuide
Synonyms: potassium trifluoro(4-methylphenyl)boranuide
Molecular Weight (g/mol): 198.04
Molecular Formula: C7H7BF3K
InChi Key: KRWDYXJWQBTBAH-UHFFFAOYSA-N
SMILES: [K+].CC1=CC=C(C=C1)[B-](F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)