accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21658-95-7

IUPAC Name: bis(propan-2-yl) (cyanomethyl)phosphonate
Synonyms: diisopropyl cyanomethylphosphonate
Molecular Weight (g/mol): 205.19
Molecular Formula: C8H16NO3P
InChi Key: MQDIIJHQSUNBKA-UHFFFAOYSA-N
SMILES: CC(C)OP(=O)(CC#N)OC(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results