accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2166-31-6

IUPAC Name: 6-phenyl-2,3-dihydropyridazin-3-one
Synonyms: 6-phenyl-2H-pyridazin-3-one
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: IJUIPRDMWWBTTQ-UHFFFAOYSA-N
SMILES: O=C1NN=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results