accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21662-09-9

IUPAC Name: (4E,6E)-deca-4,6-dien-1-ol
Synonyms: cis-4-decenal
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: FJHNCSRGAAVOQK-YTXTXJHMSA-N
SMILES: CCC\C=C\C=C\CCCO
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)