accessibility menu, dialog, popup

UserName

Viewing profile as user:
2H2H2H2H2H2H2H2H2H2H2H2H2H2H

CAS RN 21666-38-6

IUPAC Name: (²H₁₄)hexane
Synonyms: (²H₁₄)hexane
Molecular Weight (g/mol): 100.26
Molecular Formula: C6H14
InChi Key: VLKZOEOYAKHREP-ZLKPZJALSA-N
SMILES: [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results