accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3CH3H3COOH3CCH3H3C

CAS RN 2167-23-9

IUPAC Name: 2,2-bis(tert-butylperoxy)butane
Synonyms: 2,2-bis(tert-butylperoxy)butane
Molecular Weight (g/mol): 234.34
Molecular Formula: C12H26O4
InChi Key: HQOVXPHOJANJBR-UHFFFAOYSA-N
SMILES: CCC(C)(OOC(C)(C)C)OOC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)