accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21678-37-5

IUPAC Name: N,N,2-trimethylpropanamide
Synonyms: N,N,2-trimethylpropanamide
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: GXMIHVHJTLPVKL-UHFFFAOYSA-N
SMILES: CC(C)C(=O)N(C)C
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)