accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21679-46-9

IUPAC Name: 2,4,8,10,13,15-hexamethyl-1λ³,5λ³,7λ³,11λ³,12λ³,16λ³-hexaoxa-6-cobaltaspiro[5.5⁶.5⁶]hexadeca-1,4,7,10,12,15-hexaene-6,6,6-tris(ylium)-3,9,14-triide
Synonyms: 2,4,8,10,13,15-hexamethyl-1λ³,5λ³,7λ³,11λ³,12λ³,16λ³-hexaoxa-6-cobaltaspiro[5.5⁶.5⁶]hexadeca-1,4,7,10,12,15-hexaene-6,6,6-tris(ylium)-3,9,14-triide
Molecular Weight (g/mol): 356.26
Molecular Formula: C15H21CoO6
InChi Key: VACIXPOMJJIDBM-UHFFFAOYSA-N
SMILES: CC1=[O][Co+3]234([O]=C(C)[CH-]1)[O]=C(C)[CH-]C(C)=[O]2.CC([CH-]C(C)=[O]3)=[O]4
Quantity 
  • (3)
  • (4)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)