accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 216955-61-2

IUPAC Name: 2-{1-[(tert-butoxy)carbonyl]piperidin-4-yl}-4-methyl-1,3-thiazole-5-carboxylic acid
Synonyms: 2-[1-(tert-butoxycarbonyl)piperidin-4-yl]-4-methyl-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 326.41
Molecular Formula: C15H22N2O4S
InChi Key: XDKKXEMKXSGKOR-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1CCN(CC1)C(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results