CAS RN 217-59-4
Triphenylene, 98%
CAS: 217-59-4 Molecular Formula: C18H12 Molecular Weight (g/mol): 228.29 MDL Number: MFCD00001108 InChI Key: SLGBZMMZGDRARJ-UHFFFAOYSA-N Synonym: 9,10-benzophenanthrene,isochrysene,9,10-benzphenanthrene,benzo l phenanthrene,1,2,3,4-dibenznaphthalene,benzophenanthrene,1,2:3,4-dibenznaphthalene,ccris 1301,triphenylen-1-yl PubChem CID: 9170 ChEBI: CHEBI:33080 IUPAC Name: triphenylene SMILES: C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21
Triphenylene, 98%
CAS: 217-59-4 Molecular Formula: C18H12 Molecular Weight (g/mol): 228.29 MDL Number: MFCD00001108 InChI Key: SLGBZMMZGDRARJ-UHFFFAOYSA-N Synonym: 9,10-benzophenanthrene,isochrysene,9,10-benzphenanthrene,benzo l phenanthrene,1,2,3,4-dibenznaphthalene,benzophenanthrene,1,2:3,4-dibenznaphthalene,ccris 1301,triphenylen-1-yl PubChem CID: 9170 ChEBI: CHEBI:33080 IUPAC Name: triphenylene SMILES: C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21
Triphenylene 96.0+%, TCI America™
CAS: 217-59-4 Molecular Formula: C18H12 Molecular Weight (g/mol): 228.29 MDL Number: MFCD00001108 InChI Key: SLGBZMMZGDRARJ-UHFFFAOYSA-N Synonym: 9,10-benzophenanthrene,isochrysene,9,10-benzphenanthrene,benzo l phenanthrene,1,2,3,4-dibenznaphthalene,benzophenanthrene,1,2:3,4-dibenznaphthalene,ccris 1301,triphenylen-1-yl PubChem CID: 9170 ChEBI: CHEBI:33080 IUPAC Name: triphenylene SMILES: C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21
Triphenylene (purified by sublimation) 98.0+%, TCI America™
CAS: 217-59-4 Molecular Formula: C18H12 Molecular Weight (g/mol): 228.29 MDL Number: MFCD00001108 InChI Key: SLGBZMMZGDRARJ-UHFFFAOYSA-N Synonym: 9,10-benzophenanthrene,isochrysene,9,10-benzphenanthrene,benzo l phenanthrene,1,2,3,4-dibenznaphthalene,benzophenanthrene,1,2:3,4-dibenznaphthalene,ccris 1301,triphenylen-1-yl PubChem CID: 9170 ChEBI: CHEBI:33080 IUPAC Name: triphenylene SMILES: C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21
Sigma Aldrich Triphenylene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Triphenylene, SPEX CertiPrep™
CAS: 217-59-4 Molecular Formula: C18H12 Molecular Weight (g/mol): 228.29 MDL Number: MFCD00001108 InChI Key: SLGBZMMZGDRARJ-UHFFFAOYSA-N PubChem CID: 9170 ChEBI: CHEBI:33080 IUPAC Name: triphenylene SMILES: C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21