CAS RN 21702-79-4
IUPAC Name:
methyl 3,5-dibromo-2-hydroxybenzoate
Synonyms:
methyl 3,5-dibromo-2-hydroxybenzoate
Molecular Weight (g/mol):
309.94
Molecular Formula:
C8H6Br2O3
InChi Key:
KISQISIKTRRMOX-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Br)=CC(Br)=C1O
1
–
2
of
2
results
1
Promotions Available
Methyl 3,5-dibromo-2-hydroxybenzoate, 98%
Methyl 3,5-dibromo-2-hydroxybenzoate, 98%
CAS: 21702-79-4 Molecular Formula: C8H6Br2O3 Molecular Weight (g/mol): 309.94 MDL Number: MFCD00017534 InChI Key: KISQISIKTRRMOX-UHFFFAOYSA-N PubChem CID: 624217 IUPAC Name: methyl 3,5-dibromo-2-hydroxybenzoate SMILES: COC(=O)C1=CC(Br)=CC(Br)=C1O
2
Sigma Aldrich Methyl 3,5-dibromo-2-hydroxybenzoate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.