accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FN

CAS RN 21717-96-4

IUPAC Name: 5-fluoropyridin-2-amine
Synonyms: 5-fluoropyridin-2-amine
Molecular Weight (g/mol): 112.11
Molecular Formula: C5H5FN2
InChi Key: YJTXQLYMECWULH-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C=N1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)