accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2174-16-5

IUPAC Name: 2-[bis(2-hydroxyethyl)amino]ethan-1-ol; 2-hydroxybenzoic acid
Synonyms: tea-salicylate trolamine salicylate
Molecular Weight (g/mol): 287.31
Molecular Formula: C13H21NO6
InChi Key: UEVAMYPIMMOEFW-UHFFFAOYSA-N
SMILES: OCCN(CCO)CCO.OC(=O)C1=CC=CC=C1O
Grade 
  • (6)
Percent Purity 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (6)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)