accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 2175-91-9

IUPAC Name: 5-(propan-2-ylidene)cyclopenta-1,3-diene
Synonyms: 5-(propan-2-ylidene)cyclopenta-1,3-diene
Molecular Weight (g/mol): 106.17
Molecular Formula: C8H10
InChi Key: WXACXMWYHXOSIX-UHFFFAOYSA-N
SMILES: CC(C)=C1C=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (3)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)