accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2177-70-0

H3CCH2OO

CAS RN 2177-70-0

IUPAC Name: phenyl 2-methylprop-2-enoate
Synonyms: phenyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: QIWKUEJZZCOPFV-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)