accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSSCH3

CAS RN 2179-60-4

IUPAC Name: 1-(methyldisulfanyl)propane
Synonyms: 1-(methyldisulfanyl)propane
Molecular Weight (g/mol): 122.24
Molecular Formula: C4H10S2
InChi Key: PUCHCUYBORIUSM-UHFFFAOYSA-N
SMILES: CCCSSC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)