accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21797-13-7

IUPAC Name: palladium(2+) tetrakis(acetonitrile) bis(tetrafluoroboranuide)
Synonyms: palladium(2+) tetrakis(acetonitrile) ditetrafluoroborate
Molecular Weight (g/mol): 444.24
Molecular Formula: C8H12B2F8N4Pd
InChi Key: YWMRPVUMBTVUEX-UHFFFAOYSA-N
SMILES: [Pd++].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F.F[B-](F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)