accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 218131-32-9

IUPAC Name: 3-[(1H-imidazol-1-yl)methyl]benzoic acid hydrochloride
Synonyms: 3-(imidazol-1-ylmethyl)benzoic acid hydrochloride
Molecular Weight (g/mol): 238.67
Molecular Formula: C11H11ClN2O2
InChi Key: YTCNDYWVEAFCMT-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=CC(CN2C=CN=C2)=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results