CAS RN 218151-78-1
IUPAC Name:
2,3-dimethyl-1-propyl-1H-imidazol-3-ium iodide
Synonyms:
1,2-dimethyl-3-propylimidazol-1-ium iodide
Molecular Weight (g/mol):
266.13
Molecular Formula:
C8H15IN2
InChi Key:
ISHFYECQSXFODS-UHFFFAOYSA-M
SMILES:
[I-].CCCN1C=C[N+](C)=C1C
1
–
1
of
1
results
1
1,2-Dimethyl-3-propylimidazolium Iodide 98.0+%, TCI America™
CAS: 218151-78-1 Molecular Formula: C8H15IN2 Molecular Weight (g/mol): 266.126 InChI Key: ISHFYECQSXFODS-UHFFFAOYSA-M PubChem CID: 11459786 IUPAC Name: 1,2-dimethyl-3-propylimidazol-1-ium;iodide SMILES: CCCN1C=C[N+](=C1C)C.[I-]