accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2O

CAS RN 2182-55-0

IUPAC Name: (ethenyloxy)cyclohexane
Synonyms: (ethenyloxy)cyclohexane
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: VGIYPVFBQRUBDD-UHFFFAOYSA-N
SMILES: C=COC1CCCCC1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)