accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2182-66-3

IUPAC Name: (acetyloxy)dimethylsilyl acetate
Synonyms: (acetyloxy)dimethylsilyl acetate
Molecular Weight (g/mol): 176.24
Molecular Formula: C6H12O4Si
InChi Key: RQVFGTYFBUVGOP-UHFFFAOYSA-N
SMILES: CC(=O)O[Si](C)(C)OC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results