accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOPOOO

CAS RN 2183-17-7

IUPAC Name: disodium naphthalen-1-yl phosphate
Synonyms: disodium naphthalen-1-yl phosphate
Molecular Weight (g/mol): 268.12
Molecular Formula: C10H7Na2O4P
InChi Key: QYURIFWAOPAPAJ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results