accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOOCl

CAS RN 21854-95-5

IUPAC Name: bis(2-chlorophenyl)ethane-1,2-dione
Synonyms: bis(2-chlorophenyl)ethane-1,2-dione
Molecular Weight (g/mol): 279.12
Molecular Formula: C14H8Cl2O2
InChi Key: VOSNNSVWVJFJCR-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(=O)C(=O)C1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results