accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2CH3OHO

CAS RN 2190-04-7

IUPAC Name: acetic acid; octadecan-1-amine
Synonyms: acetic acid; octadecylamine
Molecular Weight (g/mol): 329.57
Molecular Formula: C20H43NO2
InChi Key: UPHWVVKYDQHTCF-UHFFFAOYSA-N
SMILES: CC(O)=O.CCCCCCCCCCCCCCCCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results