accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21900-24-3

IUPAC Name: 4-chloro-3-methylbenzoyl chloride
Synonyms: 4-chloro-3-methylbenzoyl chloride
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: NECBSSKNQDXVKN-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC(=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results