accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FClO

CAS RN 21900-39-0

IUPAC Name: 5-fluoro-2-methylbenzoyl chloride
Synonyms: 5-fluoro-2-methylbenzoyl chloride
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: HBAQRSDZSVESSF-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results