accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OCl

CAS RN 21900-62-9

IUPAC Name: 4-(propan-2-yl)benzoyl chloride
Synonyms: 4-isopropylbenzoyl chloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: LBSYWDTVBUZMCM-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results