accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNOO

CAS RN 21901-18-8

IUPAC Name: 4-methyl-3-nitro-1,2-dihydropyridin-2-one
Synonyms: 4-methyl-3-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: HZCWTTHQRMHIIE-UHFFFAOYSA-N
SMILES: CC1=C(C(=O)NC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)