accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBr

CAS RN 21906-32-1

IUPAC Name: 1-(3-bromophenyl)propan-2-one
Synonyms: 1-(3-bromophenyl)propan-2-one
Molecular Weight (g/mol): 213.07
Molecular Formula: C9H9BrO
InChi Key: FQIZFAJMBXZVOL-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results