accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFOCs

CAS RN 21907-50-6

IUPAC Name: caesium(1+) trifluoroacetate
Synonyms: caesium(1+) trifluoroacetate
Molecular Weight (g/mol): 245.92
Molecular Formula: C2CsF3O2
InChi Key: RJYSYRSELCQCSO-UHFFFAOYSA-M
SMILES: [Cs+].[O-]C(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results