accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClHClHOONNCl

CAS RN 2192-20-3

IUPAC Name: 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)ethan-1-ol dihydrochloride
Synonyms: hydroxyzine dihydrochloride
Molecular Weight (g/mol): 447.83
Molecular Formula: C21H29Cl3N2O2
InChi Key: ANOMHKZSQFYSBR-UHFFFAOYNA-N
SMILES: Cl.Cl.OCCOCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
Grade 
  • (8)
Percent Purity 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (3)
  • (8)
  • (8)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)